Anti-NR1I2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA318336EA 666 EUR
ORIGTA318336
Anti-NR1I2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:4000; Immunochemistry: 3.75 µg/ml; Western Blot: 1-3 µg/ml

Type: Primary
Antigen: NR1I2 (nuclear receptor subfamily 1, group I, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR