Anti-ALPL Mouse Monoclonal Antibody

Proveedor: OriGene
ORIGTA318268EA 666 EUR
ORIGTA318268
Anti-ALPL Mouse Monoclonal Antibody
Anticuerpos
Type: Primary
Antigen: ALPL (alkaline phosphatase, liver/bone/kidney)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR