Anti-UCP2 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA318227EA 666 EUR
ORIGTA318227
Anti-UCP2 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:32000; Immunochemistry: 3.75 µg/ml; Western Blot: 1:32000 1-3 µg/ml

Type: Primary
Antigen: UCP2 (uncoupling protein 2 (mitochondrial, proton carrier))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR