Anti-CTSK Rabbit Polyclonal Antibody

Proveedor: OriGene
TA318065
ORIGTA318065EA 834 EUR
ORIGTA318065
Anti-CTSK Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml; Immunoprecipitation; Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CTSK (cathepsin K)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR