Anti-B2M Mouse Monoclonal Antibody [clone: 2213]

Proveedor: OriGene
ORIGTA318002EA 666 EUR
ORIGTA318002
Anti-B2M Mouse Monoclonal Antibody [clone: 2213]
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml

Type: Primary
Antigen: B2M (beta-2-microglobulin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR