Anti-ADAMTS1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317919EA 680 EUR
ORIGTA317919
Anti-ADAMTS1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1.3 µg/ml

Type: Primary
Antigen: ADAMTS1 (ADAM metallopeptidase with thrombospondin type 1 motif, 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR