Anti-IL21 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317844EA 793 EUR
ORIGTA317844
Anti-IL21 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: IL21 (interleukin 21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR