Anti-GNAS Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317706EA 666 EUR
ORIGTA317706
Anti-GNAS Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:000-1:1000; Immunochemistry: 20 µg/ml

Type: Primary
Antigen: GNAS (GNAS complex locus)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR