Anti-PLA2G3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317693EA 666 EUR
ORIGTA317693
Anti-PLA2G3 Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: PLA2G3 (phospholipase A2, group III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR