Anti-CCR4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317643EA 666 EUR
ORIGTA317643
Anti-CCR4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 2-3 µg/ml

Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR