Anti-AGTR2 Rabbit Polyclonal Antibody

Proveedor: OriGene
TA317626
ORIGTA317626EA 680 EUR
ORIGTA317626
Anti-AGTR2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: AGTR2 (angiotensin II receptor, type 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR