Anti-AVPR1B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317556EA 680 EUR
ORIGTA317556
Anti-AVPR1B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 4 µg/ml

Type: Primary
Antigen: AVPR1B (arginine vasopressin receptor 1B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR