Anti-UBC Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317482EA 666 EUR
ORIGTA317482
Anti-UBC Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:1000; Immunochemistry: 10 µg/ml; Western Blot: 1:100-1:500

Type: Primary
Antigen: UBC (ubiquitin C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR