Anti-TRPV4 Rabbit Polyclonal Antibody

Proveedor: OriGene
TA317467
ORIGTA317467EA 680 EUR
ORIGTA317467
Anti-TRPV4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 5-7.5 µg/ml

Type: Primary
Antigen: TRPV4 (transient receptor potential cation channel, subfamily V, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR