Anti-SLC5A4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317341EA 680 EUR
ORIGTA317341
Anti-SLC5A4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: SLC5A4 (solute carrier family 5 (glucose activated ion channel), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR