Anti-SLC17A7 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA317309EA 666 EUR
ORIGTA317309
Anti-SLC17A7 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 3.75 µg/ml

Type: Primary
Antigen: SLC17A7 (solute carrier family 17 (vesicular glutamate transporter), member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR