Anti-SLC11A2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317307EA 666 EUR
ORIGTA317307
Anti-SLC11A2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 15 µg/ml

Type: Primary
Antigen: SLC11A2 (solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR