Anti-SIK2 Mouse Monoclonal Antibody [clone: S15G10]

Proveedor: OriGene
ORIGTA317304EA 666 EUR
ORIGTA317304
Anti-SIK2 Mouse Monoclonal Antibody [clone: S15G10]
Anticuerpos
Recommended Dilutions: Immunochemistry: 10 µg/ml

Type: Primary
Antigen: SIK2 (salt-inducible kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR