Anti-SCNN1G Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317268EA 680 EUR
ORIGTA317268
Anti-SCNN1G Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 1 µg/ml

Type: Primary
Antigen: SCNN1G (sodium channel, non-voltage-gated 1, gamma subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR