Anti-PCSK6 Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA317144EA 666 EUR
ORIGTA317144
Anti-PCSK6 Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:8000; Immunochemistry: 3.75 µg/ml

Type: Primary
Antigen: PCSK6 (proprotein convertase subtilisin/kexin type 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR