Anti-NR2E3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317090EA 666 EUR
ORIGTA317090
Anti-NR2E3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 7 µg/ml

Type: Primary
Antigen: NR2E3 (nuclear receptor subfamily 2, group E, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR