Anti-MTSS1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317043EA 666 EUR
ORIGTA317043
Anti-MTSS1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 10 µg/ml; Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MTSS1 (metastasis suppressor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR