Anti-MRGPRX2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA317038EA 666 EUR
ORIGTA317038
Anti-MRGPRX2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: MRGPRX2 (MAS-related GPR, member X2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR