Anti-KCNMA1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316944EA 680 EUR
ORIGTA316944
Anti-KCNMA1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: KCNMA1 (potassium large conductance calcium-activated channel, subfamily M, alpha member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR