Anti-INPP5J Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316924EA 666 EUR
ORIGTA316924
Anti-INPP5J Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 6 µg/ml

Type: Primary
Antigen: INPP5J (inositol polyphosphate-5-phosphatase J)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR