Anti-IGFBP5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316916EA 680 EUR
ORIGTA316916
Anti-IGFBP5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml; Western Blot: 1:200

Type: Primary
Antigen: IGFBP5 (insulin-like growth factor binding protein 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR