Anti-HTR6 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316906EA 666 EUR
ORIGTA316906
Anti-HTR6 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 20 µg/ml

Type: Primary
Antigen: HTR6 (5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR