Anti-HNF4G Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316895EA 666 EUR
ORIGTA316895
Anti-HNF4G Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 16-32 µg/ml

Type: Primary
Antigen: HNF4G (hepatocyte nuclear factor 4, gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR