Anti-HFE2 Mouse Monoclonal Antibody [clone: 1C12]

Proveedor: OriGene
ORIGTA316883EA 867 EUR
ORIGTA316883
Anti-HFE2 Mouse Monoclonal Antibody [clone: 1C12]
Anticuerpos
Recommended Dilutions: Immunochemistry: 5 µg/ml;

Type: Primary
Antigen: HFE2 (hemochromatosis type 2 (juvenile))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR