Anti-HCAR1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316878EA 666 EUR
ORIGTA316878
Anti-HCAR1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 3 µg/ml

Type: Primary
Antigen: HCAR1 (hydroxycarboxylic acid receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR