Anti-GRPR Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316872EA 666 EUR
ORIGTA316872
Anti-GRPR Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 4 µg/ml;

Type: Primary
Antigen: GRPR (gastrin-releasing peptide receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR