Anti-GPR78 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316852EA 666 EUR
ORIGTA316852
Anti-GPR78 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 3-6 µg/ml

Type: Primary
Antigen: GPR78 (G protein-coupled receptor 78)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR