Anti-CLTC Goat Polyclonal Antibody

Proveedor: OriGene
ORIGTA316635EA 666 EUR
ORIGTA316635
Anti-CLTC Goat Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 3.75 µg/ml

Type: Primary
Antigen: CLTC (clathrin, heavy chain (Hc))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR