Anti-C4B Chicken Polyclonal Antibody

Proveedor: OriGene
TA316579
ORIGTA316579EA 680 EUR
ORIGTA316579
Anti-C4B Chicken Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA Flo (0.1 µg/1E6 cells; Immunochemistry: 20 µg/ml

Type: Primary
Antigen: C4B (complement component 4B (Chido blood group))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR