Anti-NT5C1B Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316490EA 552 EUR
ORIGTA316490
Anti-NT5C1B Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: NT5C1B (5'-nucleotidase, cytosolic IB)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR