Anti-NKX2-4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316482EA 552 EUR
ORIGTA316482
Anti-NKX2-4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: NKX2-4 (NK2 homeobox 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR