Anti-OR10C1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316475EA 552 EUR
ORIGTA316475
Anti-OR10C1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: OR10C1 (olfactory receptor, family 10, subfamily C, member 1 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR