Anti-OR14J1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316474EA 564 EUR
ORIGTA316474
Anti-OR14J1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: OR14J1 (olfactory receptor, family 14, subfamily J, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR