Anti-OR51D1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316405EA 564 EUR
ORIGTA316405
Anti-OR51D1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: OR51D1 (olfactory receptor, family 51, subfamily D, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR