Anti-OR2AG2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316343EA 552 EUR
ORIGTA316343
Anti-OR2AG2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:1000

Type: Primary
Antigen: OR2AG2 (olfactory receptor, family 2, subfamily AG, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR