Anti-VN1R5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316336EA 543 EUR
ORIGTA316336
Anti-VN1R5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: VN1R5 (vomeronasal 1 receptor 5 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR