Anti-RIMKLA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316319EA 552 EUR
ORIGTA316319
Anti-RIMKLA Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: RIMKLA (ribosomal modification protein rimK-like family member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR