Anti-OR8B4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316301EA 552 EUR
ORIGTA316301
Anti-OR8B4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:5000

Type: Primary
Antigen: OR8B4 (olfactory receptor, family 8, subfamily B, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR