Anti-CYP4Z1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316216EA 532 EUR
ORIGTA316216
Anti-CYP4Z1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:20000

Type: Primary
Antigen: CYP4Z1 (cytochrome P450, family 4, subfamily Z, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR