Anti-PAQR7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316189EA 552 EUR
ORIGTA316189
Anti-PAQR7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: PAQR7 (progestin and adipoQ receptor family member VII)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR