Anti-GDF7 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316168EA 532 EUR
ORIGTA316168
Anti-GDF7 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: GDF7 (growth differentiation factor 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR