Anti-SLC24A4 Rabbit Polyclonal Antibody

Proveedor: OriGene
TA316067
ORIGTA316067EA 564 EUR
ORIGTA316067
Anti-SLC24A4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:40000

Type: Primary
Antigen: SLC24A4 (solute carrier family 24 (sodium/potassium/calcium exchanger), member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR