Anti-CYP2R1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316046EA 543 EUR
ORIGTA316046
Anti-CYP2R1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:40000

Type: Primary
Antigen: CYP2R1 (cytochrome P450, family 2, subfamily R, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR