Anti-GBP4 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA316020EA 532 EUR
ORIGTA316020
Anti-GBP4 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: GBP4 (guanylate binding protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR