Anti-CCDC102A Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA315992EA 552 EUR
ORIGTA315992
Anti-CCDC102A Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:1000

Type: Primary
Antigen: CCDC102A (coiled-coil domain containing 102A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR